siRNA Details
Gene Symbol: TBL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012453.2 | NP_036585.1 | 1 | 250 | ||
XM_011516027.2 | XP_011514329.1 | 1 | 143 | |||
GenBank mRNA | AF056183.1 | - | 1 | 78 | ||
AF097484.1 | - | 1 | 135 | |||
AK000536.1 | - | 1 | 143 | |||
AK074619.1 | - | 1 | 146 | |||
AK130791.1 | - | 1 | 143 | |||
AK302277.1 | - | 1 | 166 | |||
AK314499.1 | - | 1 | 143 | |||
AL080162.1 | - | 1 | 210 | |||
AY358518.1 | - | 1 | 162 | |||
BC012938.1 | - | 1 | 127 | 4453977 | ||
BC047354.1 | - | 1 | 188 | |||
BX420755.2 | - | 1 | 251 | |||
KU178597.1 | - | 1 | 116 | |||
KU178598.1 | - | 1 | 116 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
