siRNA Details
Gene Symbol: PHACTR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100164.1 | NP_001093634.1 | 11 | 1970 | ||
NM_001100165.1 | NP_001093635.1 | 10 | 1730 | |||
NM_001100166.1 | NP_001093636.1 | 10 | 1628 | |||
NM_014721.2 | NP_055536.2 | 11 | 1868 | |||
GenBank mRNA | AB014580.1 | - | 11 | 1785 | ||
AK297799.1 | - | 11 | 1775 | |||
AK298263.1 | - | 11 | 1970 | |||
AK300221.1 | - | 10 | 1623 | |||
BC150630.1 | - | 11 | 1775 | 9057000 | ||
BC152416.1 | - | 11 | 1785 | 8862521 | ||
CR749345.1 | - | 10 | 1668 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
