siRNA Details
Gene Symbol: ZNF853 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017560.1 | NP_060030.1 | 3 | 1607 | ||
XM_011515438.2 | XP_011513740.1 | 4 | 1769 | |||
XM_011515439.2 | XP_011513741.1 | 4 | 1773 | |||
XM_017012371.1 | XP_016867860.1 | 3 | 1887 | |||
GenBank mRNA | AL133055.1 | - | 1 | 384 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
