siRNA Details
Gene Symbol: SLN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003063.2 | NP_003054.1 | 2 | 572 | ||
GenBank mRNA | BC005261.1 | - | 2 | 567 | 3934241 | |
BC094685.1 | - | 2 | 554 | 6203008 | ||
BP233121.1 | - | 2 | 572 | |||
F30315.1 | - | 1 | 186 | |||
U96094.1 | - | 2 | 566 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
