siRNA Details
Gene Symbol: SNX7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015976.4 | NP_057060.2 | 2 | 309 | ||
NM_152238.3 | NP_689424.2 | 2 | 309 | |||
NR_033716.1 | 3 | 429 | ||||
XM_006710677.2 | XP_006710740.1 | 2 | 326 | |||
XM_006710678.2 | XP_006710741.1 | 3 | 425 | |||
XM_011541564.2 | XP_011539866.1 | 3 | 365 | |||
XM_017001425.1 | XP_016856914.1 | 2 | 287 | |||
XM_017001426.1 | XP_016856915.1 | 3 | 424 | |||
XM_017001427.1 | XP_016856916.1 | 3 | 699 | |||
XM_017001428.1 | XP_016856917.1 | 2 | 174 | |||
GenBank mRNA | AF121857.1 | - | 2 | 152 | ||
AK025031.1 | - | 2 | 266 | |||
AK223229.1 | - | 2 | 168 | |||
AK293018.1 | - | 2 | 237 | |||
AK298211.1 | - | 2 | 155 | |||
AL049989.1 | - | 2 | 166 | |||
AL548028.3 | - | 2 | 310 | |||
BC010349.1 | - | 2 | 123 | 4093672 | ||
BC018105.1 | - | 3 | 247 | 3849824 | ||
BI856196.1 | - | 2 | 330 | |||
BT006747.1 | - | 1 | 65 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
