siRNA Details
Gene Symbol: MUC17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040105.1 | NP_001035194.1 | 7 | 12844 | ||
GenBank mRNA | AF430017.1 | - | 5 | 2401 | ||
AJ606307.1 | - | 7 | 12844 | |||
AK026404.1 | - | 5 | 1515 | |||
BC126315.1 | - | 5 | 2986 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
