siRNA Details
Gene Symbol: MUC17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040105.1 | NP_001035194.1 | 7 | 12867 | ||
GenBank mRNA | AJ606307.1 | - | 7 | 12867 | ||
BC126315.1 | - | 5 | 3009 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
