siRNA Details
Gene Symbol: FOXI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012188.4 | NP_036320.2 | 2 | 1747 | ||
NM_144769.2 | NP_658982.1 | 2 | 1462 | |||
GenBank mRNA | BC029778.1 | - | 2 | 1462 | 5185923 | |
L13203.1 | - | 2 | 1699 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
