siRNA Details
Gene Symbol: ZNF608 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020747.2 | NP_065798.2 | 4 | 3719 | ||
XM_005272037.2 | XP_005272094.1 | 5 | 4481 | |||
XM_005272038.1 | XP_005272095.1 | 5 | 4071 | |||
XM_005272039.2 | XP_005272096.1 | 5 | 4481 | |||
XM_005272040.3 | XP_005272097.1 | 4 | 2967 | |||
XM_011543520.2 | XP_011541822.1 | 5 | 4481 | |||
XM_011543522.2 | XP_011541824.1 | 4 | 3137 | |||
XM_011543523.2 | XP_011541825.1 | 3 | 2579 | |||
XM_017009674.1 | XP_016865163.1 | 5 | 4481 | |||
GenBank mRNA | AB033107.2 | - | 4 | 3720 | ||
BC151226.1 | - | 4 | 3720 | 9007221 | ||
CR749624.1 | - | 4 | 2800 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
