siRNA Details
Gene Symbol: ZNF608 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020747.2 | NP_065798.2 | 9 | 5168 | ||
XM_005272037.2 | XP_005272094.1 | 10 | 5930 | |||
XM_005272038.1 | XP_005272095.1 | 10 | 5520 | |||
XM_005272039.2 | XP_005272096.1 | 11 | 5972 | |||
XM_005272040.3 | XP_005272097.1 | 9 | 4416 | |||
XM_011543522.2 | XP_011541824.1 | 9 | 4586 | |||
XM_011543523.2 | XP_011541825.1 | 8 | 4028 | |||
GenBank mRNA | AB033107.2 | - | 9 | 5169 | ||
AL117587.1 | - | 6 | 1058 | |||
BC038370.1 | - | 5 | 1230 | |||
BC045828.2 | - | 2 | 557 | |||
BC131507.1 | - | 8 | 2570 | |||
BC151226.1 | - | 9 | 5169 | 9007221 | ||
CR749624.1 | - | 9 | 4251 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
