siRNA Details
Gene Symbol: PSG11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113410.1 | NP_001106881.1 | 4 | 722 | ||
NM_002785.2 | NP_002776.3 | 5 | 1088 | |||
NM_203287.1 | NP_976032.2 | 4 | 722 | |||
GenBank mRNA | BC020711.1 | - | 4 | 712 | 4732286 | |
BE767872.1 | - | 2 | 191 | |||
BG571293.1 | - | 3 | 431 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
