siRNA Details
Gene Symbol: CHTF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039690.3 | NP_001034779.1 | 4 | 1053 | ||
NM_001040146.3 | NP_001035236.1 | 4 | 1026 | |||
NR_033227.1 | 3 | 995 | ||||
XM_011523167.1 | XP_011521469.1 | 3 | 932 | |||
XM_011523168.1 | XP_011521470.1 | 3 | 886 | |||
GenBank mRNA | AF525164.1 | - | 4 | 1033 | ||
AK000407.1 | - | 4 | 1033 | |||
AL833902.1 | - | 1 | 612 | |||
BC018700.2 | - | 4 | 1000 | 3504183 | ||
BC069059.1 | - | 3 | 916 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
