siRNA Details
Gene Symbol: DAZ4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005375.2 | NP_001005375.1 | 21, 22 | 1983 | ||
NM_020420.3 | NP_065153.1 | 15, 16 | 1416 | |||
XM_005262602.1 | XP_005262659.1 | 27, 28 | 2354 | |||
XM_011531509.2 | XP_011529811.1 | 27, 28 | 2441 | |||
XM_011531510.1 | XP_011529812.1 | 21, 22 | 1565 | |||
GenBank mRNA | AF248483.1 | - | 15, 16 | 1178 | ||
BC047480.1 | - | 15, 16 | 1321 | 5297459 | ||
BC047617.1 | - | 15, 16 | 1317 | 4836748 |
Gene Symbol: DAZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004081.5 | NP_004072.3 | 26, 27 | 2463 | ||
XM_011531482.2 | XP_011529784.1 | 26, 27 | 2501 | |||
XM_011531483.1 | XP_011529785.1 | 20, 21 | 1628 | |||
GenBank mRNA | AF271087.1 | - | 14, 15 | 1840 | ||
AF271088.1 | - | 18, 19 | 1443 | |||
AK310133.1 | - | 2, 3 | 234 |
Gene Symbol: DAZ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005785.2 | NP_001005785.1 | 21, 22 | 1825 | ||
NM_001005786.2 | NP_001005786.2 | 14, 15 | 1321 | |||
NM_020363.3 | NP_065096.2 | 22, 23 | 1897 | |||
GenBank mRNA | AF248480.1 | - | 22, 23 | 1809 | ||
AF414184.1 | - | 21, 22 | 1807 | |||
AK302398.1 | - | 24, 25 | 1964 | |||
AK302690.1 | - | 25, 26 | 1966 | |||
BC113006.1 | - | 21, 22 | 1633 | 40035862 | ||
U21663.1 | - | 14, 15 | 1245 |
Gene Symbol: DAZ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020364.3 | NP_065097.2 | 17, 18 | 1560 | ||
GenBank mRNA | AF248481.1 | - | 19, 20 | 1585 | ||
BC113005.1 | - | 17, 18 | 1345 | 40035860 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
