siRNA Details
Gene Symbol: DAZ4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005375.2 | NP_001005375.1 | 12, 13 | 1369 | ||
NM_020420.3 | NP_065153.1 | 7, 8 | 874 | |||
XM_005262602.1 | XP_005262659.1 | 12, 13 | 1308 | |||
XM_011531509.2 | XP_011529811.1 | 12, 13 | 1395 | |||
XM_011531510.1 | XP_011529812.1 | 6, 7 | 519 | |||
GenBank mRNA | AF248483.1 | - | 6, 7 | 564 | ||
BC047480.1 | - | 7, 8 | 779 | 5297459 | ||
BC047617.1 | - | 7, 8 | 775 | 4836748 |
Gene Symbol: DAZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004081.5 | NP_004072.3 | 17, 18 | 1849 | ||
XM_011531482.2 | XP_011529784.1 | 17, 18 | 1887 | |||
XM_011531483.1 | XP_011529785.1 | 11, 12 | 1014 | |||
GenBank mRNA | AF271087.1 | - | 5, 6 | 1226 | ||
AF271088.1 | - | 9, 10 | 829 | |||
BC114927.1 | - | 17, 18 | 1680 | 40028543 |
Gene Symbol: DAZ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005785.2 | NP_001005785.1 | 7, 8 | 851 | ||
NM_001005786.2 | NP_001005786.2 | 7, 8 | 851 | |||
NM_020363.3 | NP_065096.2 | 7, 8 | 851 | |||
GenBank mRNA | AF248480.1 | - | 7, 8 | 763 | ||
AF414184.1 | - | 7, 8 | 833 | |||
AK302398.1 | - | 7, 8 | 774 | |||
AK302690.1 | - | 6, 7 | 704 | |||
BC113006.1 | - | 7, 8 | 659 | 40035862 | ||
U21663.1 | - | 7, 8 | 775 |
Gene Symbol: DAZ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020364.3 | NP_065097.2 | 7, 8 | 874 | ||
GenBank mRNA | AF248481.1 | - | 7, 8 | 755 | ||
BC113005.1 | - | 7, 8 | 659 | 40035860 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
