siRNA Details
Gene Symbol: KIAA1586 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286274.1 | NP_001273203.1 | 3 | 2750 | ||
NM_001286275.1 | NP_001273204.1 | 4 | 2809 | |||
NM_001286276.1 | NP_001273205.1 | 4 | 2761 | |||
NM_020931.3 | NP_065982.1 | 4 | 2831 | |||
GenBank mRNA | AB046806.1 | - | 1 | 3944 | ||
BC041125.1 | - | 4 | 2775 | 5785266 | ||
CN359914.1 | - | 1 | 648 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
