siRNA Details
Gene Symbol: KIAA1586 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286274.1 | NP_001273203.1 | 3 | 647 | ||
NM_001286275.1 | NP_001273204.1 | 4 | 706 | |||
NM_001286276.1 | NP_001273205.1 | 4 | 658 | |||
NM_020931.3 | NP_065982.1 | 4 | 728 | |||
GenBank mRNA | AB046806.1 | - | 1 | 1840 | ||
AK290988.1 | - | 4 | 522 | |||
AK295519.1 | - | 3 | 453 | |||
BC029372.1 | - | 4 | 537 | |||
BC041125.1 | - | 4 | 674 | 5785266 | ||
BG028983.1 | - | 494 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
