siRNA Details
Gene Symbol: COG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032382.4 | NP_115758.3 | 6 | 2095 | ||
GenBank mRNA | AK025968.1 | - | 4 | 1217 | ||
AK056344.1 | - | 6 | 2083 | |||
AK225391.1 | - | 4 | 1217 | |||
AK302605.1 | - | 6 | 2108 | |||
BC017492.1 | - | 4 | 899 |
Gene Symbol: PDF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022341.1 | NP_071736.1 | 2 | 758 | ||
GenBank mRNA | AF239156.1 | - | 2 | 758 | ||
AF322879.1 | - | 2 | 733 | |||
BC019912.1 | - | 2 | 734 | 4994286 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
