siRNA Details
Gene Symbol: OR10A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004460.1 | NP_001004460.1 | 1 | 725 | ||
GenBank mRNA | BC136955.1 | - | 1 | 787 | 9020947 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
