siRNA Details
Gene Symbol: OR2AG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004489.2 | NP_001004489.1 | 1 | 118 | ||
GenBank mRNA | BC151143.1 | - | 1 | 118 | 9020805 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
