siRNA Details
Gene Symbol: MCRIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138418.2 | NP_612427.2 | 5 | 818 | ||
GenBank mRNA | AF386504.1 | - | 4 | 691 | ||
AK054811.1 | - | 3 | 2214 | |||
BC001912.1 | - | 5 | 818 | 3546959 | ||
BC007346.2 | - | 4 | 610 | |||
BC111391.1 | - | 5 | 721 | 6457301 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
