siRNA Details
Gene Symbol: KIAA0232 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100590.1 | NP_001094060.1 | 6 | 2957 | ||
NM_014743.2 | NP_055558.2 | 7 | 3041 | |||
XM_005248041.4 | XP_005248098.1 | 4 | 2516 | |||
XM_011513598.2 | XP_011511900.1 | 7 | 2694 | |||
XM_017008860.1 | XP_016864349.1 | 6 | 3987 | |||
XM_017008861.1 | XP_016864350.1 | 5 | 2704 | |||
XM_017008862.1 | XP_016864351.1 | 6 | 3342 | |||
XM_017008863.1 | XP_016864352.1 | 5 | 2704 | |||
GenBank mRNA | BC150286.1 | - | 7 | 3033 | 8860111 | |
D86985.3 | - | 7 | 3033 | |||
EF553521.1 | - | 6 | 2898 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
