siRNA Details
Gene Symbol: SEMA3E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178129.1 | NP_001171600.1 | 17 | 3179 | ||
NM_012431.2 | NP_036563.1 | 17 | 3815 | |||
GenBank mRNA | AB002329.1 | - | 17 | 3660 | ||
BC140706.1 | - | 17 | 3350 | 9021574 | ||
BC144338.1 | - | 17 | 3350 | 9052858 | ||
BC152458.1 | - | 17 | 3672 | 8862593 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
