siRNA Details
Gene Symbol: KRTAP5-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021046.2 | NP_066384.2 | 1 | 580 | ||
GenBank mRNA | AB126077.1 | - | 1 | 580 | ||
AY360461.1 | - | 1 | 549 | |||
BC119736.1 | - | 4 | 633 |
Gene Symbol: KRTAP5-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012708.2 | NP_001012726.1 | 1 | 780 | ||
GenBank mRNA | AB126072.1 | - | 1 | 780 | ||
AJ628246.1 | - | 1 | 490 | |||
AY597812.1 | - | 1 | 702 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
