siRNA Details
Gene Symbol: PCDHGA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018913.2 | NP_061736.1 | 4 | 3237 | ||
GenBank mRNA | AF152319.1 | - | 4 | 3237 |
Gene Symbol: PCDHGA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018912.2 | NP_061735.1 | 4 | 3222 | ||
GenBank mRNA | AF152318.1 | - | 4 | 3222 |
Gene Symbol: PCDHGA11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018914.2 | NP_061737.1 | 4 | 3492 | ||
NM_032092.1 | NP_114481.1 | 4 | 2937 | |||
GenBank mRNA | AF152320.1 | - | 4 | 3234 | ||
BC136781.1 | - | 4 | 3267 | 9020771 | ||
BC171774.1 | - | 4 | 3267 | 9054428 |
Gene Symbol: PCDHGA12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003735.2 | NP_003726.1 | 4 | 3394 | ||
GenBank mRNA | AB011160.1 | - | 4 | 3367 | ||
AF152321.1 | - | 4 | 3225 |
Gene Symbol: PCDHGA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018915.3 | NP_061738.1 | 4 | 3437 | ||
GenBank mRNA | AF152322.1 | - | 4 | 3225 |
Gene Symbol: PCDHGA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018916.3 | NP_061739.2 | 4 | 3225 | ||
GenBank mRNA | AB208927.1 | - | 4 | 3391 | ||
AF152323.1 | - | 4 | 3225 |
Gene Symbol: PCDHGA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018917.3 | NP_061740.2 | 4 | 3398 | ||
GenBank mRNA | AF152324.1 | - | 4 | 3222 |
Gene Symbol: PCDHGA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018918.2 | NP_061741.1 | 4 | 3222 | ||
GenBank mRNA | AF152325.1 | - | 4 | 3222 |
Gene Symbol: PCDHGA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018919.2 | NP_061742.1 | 4 | 3225 | ||
GenBank mRNA | AB209178.1 | - | 4 | 3395 | ||
AF152326.1 | - | 4 | 3225 | |||
BC146651.1 | - | 4 | 3384 | 40147340 |
Gene Symbol: PCDHGA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018920.3 | NP_061743.1 | 4 | 3386 | ||
GenBank mRNA | AB208958.1 | - | 4 | 3116 | ||
AF152327.1 | - | 4 | 3225 |
Gene Symbol: PCDHGA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032088.1 | NP_114477.1 | 4 | 4123 | ||
GenBank mRNA | AF152328.1 | - | 4 | 3225 |
Gene Symbol: PCDHGA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018921.2 | NP_061744.1 | 4 | 3225 | ||
GenBank mRNA | AF152329.1 | - | 4 | 3225 |
Gene Symbol: PCDHGB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018922.2 | NP_061745.1 | 4 | 3210 | ||
GenBank mRNA | AF152330.1 | - | 4 | 3210 |
Gene Symbol: PCDHGB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018923.2 | NP_061746.1 | 4 | 3222 | ||
GenBank mRNA | AF152331.1 | - | 4 | 3222 |
Gene Symbol: PCDHGB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018924.4 | NP_061747.2 | 4 | 3347 | ||
GenBank mRNA | AF152332.1 | - | 4 | 3216 | ||
BC150172.1 | - | 4 | 3347 | 40147143 |
Gene Symbol: PCDHGB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003736.2 | NP_003727.1 | 4 | 3198 | ||
GenBank mRNA | AF152333.1 | - | 4 | 3198 |
Gene Symbol: PCDHGB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018925.2 | NP_061748.1 | 4 | 3198 | ||
GenBank mRNA | AF152334.1 | - | 4 | 3198 |
Gene Symbol: PCDHGB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018926.2 | NP_061749.1 | 4 | 3219 | ||
GenBank mRNA | AF152335.1 | - | 4 | 3219 |
Gene Symbol: PCDHGB7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018927.3 | NP_061750.1 | 4 | 3383 | ||
GenBank mRNA | AF152336.1 | - | 4 | 3216 | ||
BC051788.2 | - | 4 | 3410 | 4941167 |
Gene Symbol: PCDHGC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002588.3 | NP_002579.2 | 4 | 3346 | ||
NM_032403.2 | NP_115779.1 | 4 | 946 | |||
GenBank mRNA | AF152337.1 | - | 4 | 3231 | ||
BC004321.1 | - | 1 | 642 | |||
BC006439.1 | - | 4 | 946 | |||
BC019299.1 | - | 4 | 3345 | 2905509 | ||
BC026218.1 | - | 4 | 3335 | 4153558 | ||
BC027451.1 | - | 4 | 944 | |||
BC027470.1 | - | 2 | 706 | |||
BC063437.1 | - | 4 | 930 | |||
L11373.1 | - | 4 | 3322 |
Gene Symbol: PCDHGC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018928.2 | NP_061751.1 | 4 | 3243 | ||
GenBank mRNA | AF152338.1 | - | 4 | 3243 | ||
BC042650.1 | - | 2 | 2085 |
Gene Symbol: PCDHGC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018929.2 | NP_061752.1 | 4 | 3261 | ||
GenBank mRNA | AF152339.1 | - | 4 | 3261 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
