siRNA Details
Gene Symbol: RLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005059.3 | NP_005050.2 | 1 | 63 | ||
NM_134441.2 | NP_604390.1 | 1 | 63 | |||
GenBank mRNA | A06925.1 | - | 1 | 32 | ||
A17315.1 | - | 1 | 32 | |||
BC126415.1 | - | 1 | 63 | 8992131 | ||
BC126419.1 | - | 1 | 63 | 8992135 | ||
X00948.1 | - | 1 | 32 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
