siRNA Details
Gene Symbol: EIF1AX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001412.3 | NP_001403.1 | 7 | 3425 | ||
GenBank mRNA | AL079283.1 | - | 1 | 457 | ||
AL832930.1 | - | 1 | 1969 | |||
BC047573.1 | - | 1 | 1274 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
