siRNA Details
Gene Symbol: STOX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020225.2 | NP_064610.1 | 3 | 3080 | ||
XM_011532129.1 | XP_011530431.1 | 3 | 3204 | |||
XM_011532130.1 | XP_011530432.1 | 3 | 3204 | |||
XM_011532131.1 | XP_011530433.1 | 6 | 2335 | |||
XM_017008466.1 | XP_016863955.1 | 3 | 3204 | |||
XM_017008467.1 | XP_016863956.1 | 4 | 2241 | |||
GenBank mRNA | AB037813.1 | - | 3 | 3080 | ||
AK289840.1 | - | 3 | 1759 | |||
BC146754.1 | - | 3 | 3080 | 9007190 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
