siRNA Details
Gene Symbol: CD200R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138806.3 | NP_620161.1 | 8 | 2233 | ||
NM_170780.2 | NP_740750.1 | 7 | 2164 | |||
GenBank mRNA | AK126349.1 | - | 7 | 2164 | ||
AY358910.1 | - | 8 | 2258 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
