siRNA Details
Gene Symbol: CST9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008693.2 | NP_001008693.2 | 2 | 477 | ||
GenBank mRNA | AF494536.1 | - | 2 | 380 | ||
BC137302.1 | - | 2 | 477 | 9021299 | ||
BC137303.1 | - | 2 | 477 | 9021300 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
