siRNA Details
Gene Symbol: ISX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303508.1 | NP_001290437.1 | 5 | 2799 | ||
GenBank mRNA | AK025181.1 | - | 4 | 3088 | ||
CR749372.1 | - | 5 | 2800 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
