siRNA Details
Gene Symbol: MRVI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098579.2 | NP_001092049.2 | 12 | 1704 | ||
NM_001100163.2 | NP_001093633.1 | 13 | 1982 | |||
NM_001100167.2 | NP_001093637.1 | 13 | 1699 | |||
NM_001206880.1 | NP_001193809.1 | 12 | 1417 | |||
NM_001206881.1 | NP_001193810.1 | 11 | 1824 | |||
NM_130385.3 | NP_569056.4 | 13 | 2035 | |||
GenBank mRNA | AF081249.1 | - | 11 | 1720 | ||
AF081250.1 | - | 11 | 1448 | |||
AK127209.1 | - | 13 | 1677 | |||
AK296336.1 | - | 13 | 1799 | |||
AK300358.1 | - | 12 | 1689 | |||
AK301296.1 | - | 12 | 1028 | |||
AK304702.1 | - | 12 | 1051 | |||
BC117127.1 | - | 13 | 1572 | 40125678 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
