siRNA Details
Gene Symbol: MIR99AHG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027790.3 | 2 | 190 | |||
NR_111004.2 | 1 | 163 | ||||
NR_111005.2 | 1 | 163 | ||||
NR_136541.1 | 2 | 243 | ||||
NR_136542.1 | 2 | 243 | ||||
GenBank mRNA | AK095614.1 | - | 2 | 190 | ||
CN313455.1 | - | 1 | 163 |

More Information
Other Species Targeted

