siRNA Details
Gene Symbol: LKAAEAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007125.1 | NP_001007126.1 | 1 | 165 | ||
XM_005260200.3 | XP_005260257.3 | 2 | 95 | |||
GenBank mRNA | BC036837.1 | - | 1 | 165 | 5173095 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
