siRNA Details
Gene Symbol: GCSAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190259.1 | NP_001177188.1 | 6 | 1267 | ||
NM_001190260.1 | NP_001177189.1 | 5 | 1216 | |||
NM_152785.4 | NP_689998.1 | 6 | 1261 | |||
XM_005247272.2 | XP_005247329.1 | 6 | 1685 | |||
XM_011512627.2 | XP_011510929.1 | 6 | 1139 | |||
XM_011512628.2 | XP_011510930.1 | 6 | 1196 | |||
XM_011512629.1 | XP_011510931.1 | 6 | 1688 | |||
XM_011512630.1 | XP_011510932.1 | 6 | 1694 | |||
GenBank mRNA | AF521911.1 | - | 6 | 1248 | ||
AY212246.1 | - | 6 | 1181 | |||
BC030506.1 | - | 6 | 1129 | 4385178 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
