siRNA Details
Gene Symbol: ZMIZ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300959.1 | NP_001287888.1 | 7, 8 | 1017 | ||
NM_031449.3 | NP_113637.3 | 7, 8 | 1108 | |||
NM_174929.2 | NP_777589.2 | 6, 7 | 1047 | |||
XM_005249866.2 | XP_005249923.1 | 7, 8 | 1132 | |||
XM_005249867.4 | XP_005249924.1 | 7, 8 | 1134 | |||
XM_005249868.4 | XP_005249925.1 | 7, 8 | 2152 | |||
XM_005249869.2 | XP_005249926.1 | 7, 8 | 1132 | |||
XM_005249870.2 | XP_005249927.1 | 7, 8 | 1132 | |||
XM_005249871.2 | XP_005249928.1 | 7, 8 | 1036 | |||
XM_005249872.2 | XP_005249929.1 | 7, 8 | 1132 | |||
XM_005249873.2 | XP_005249930.1 | 7, 8 | 1036 | |||
XM_017012674.1 | XP_016868163.1 | 7, 8 | 1213 | |||
GenBank mRNA | AB067473.1 | - | 2, 3 | 270 | ||
AF357908.1 | - | 6, 7 | 985 | |||
AK090415.1 | - | 7, 8 | 1108 | |||
AY426594.1 | - | 6, 7 | 1009 | |||
BC016968.1 | - | 6, 7 | 1030 | |||
BC021924.1 | - | 1, 2 | 40 | |||
BC110435.1 | - | 7, 8 | 1002 | 5518312 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
