siRNA Details
Gene Symbol: ZMIZ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300959.1 | NP_001287888.1 | 8 | 1162 | ||
NM_031449.3 | NP_113637.3 | 9 | 1331 | |||
NM_174929.2 | NP_777589.2 | 7 | 1192 | |||
XM_005249866.2 | XP_005249923.1 | 9 | 1355 | |||
XM_005249867.4 | XP_005249924.1 | 9 | 1357 | |||
XM_005249868.4 | XP_005249925.1 | 9 | 2375 | |||
XM_005249869.2 | XP_005249926.1 | 9 | 1355 | |||
XM_005249870.2 | XP_005249927.1 | 8 | 1277 | |||
XM_005249871.2 | XP_005249928.1 | 9 | 1259 | |||
XM_005249872.2 | XP_005249929.1 | 8 | 1277 | |||
XM_005249873.2 | XP_005249930.1 | 9 | 1259 | |||
XM_011515566.1 | XP_011513868.1 | 3 | 495 | |||
XM_017012674.1 | XP_016868163.1 | 9 | 1436 | |||
GenBank mRNA | AB067473.1 | - | 3 | 415 | ||
AF357908.1 | - | 8 | 1208 | |||
AK090415.1 | - | 9 | 1331 | |||
AK090841.1 | - | 1 | 611 | |||
AL831933.1 | - | 1 | 828 | |||
AY426594.1 | - | 7 | 1154 | |||
BC016968.1 | - | 7 | 1175 | |||
BC021924.1 | - | 3 | 263 | |||
BC110435.1 | - | 8 | 1147 | 5518312 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
