siRNA Details
Gene Symbol: PPP1R14B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138689.2 | NP_619634.1 | 4 | 813 | ||
GenBank mRNA | AF318364.1 | - | 3 | 814 | ||
BC014522.2 | - | 4 | 665 | 3831227 | ||
BC071884.1 | - | 1 | 568 | |||
BC094817.1 | - | 4 | 696 | 5466876 | ||
X91195.1 | - | 4 | 798 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
