siRNA Details
Gene Symbol: LIMK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031801.1 | NP_001026971.1 | 15 | 2606 | ||
GenBank mRNA | BC013051.1 | - | 15 | 2481 | 3861412 |
Gene Symbol: PPP1R14B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138689.2 | NP_619634.1 | 4 | 902 | ||
GenBank mRNA | AF318364.1 | - | 3 | 903 | ||
BC014522.2 | - | 4 | 754 | 3831227 | ||
BC071884.1 | - | 1 | 657 | |||
BC094817.1 | - | 4 | 785 | 5466876 | ||
X91195.1 | - | 4 | 887 |

More Information
Other Species Targeted

Gene Ontology Categories:
