siRNA Details
Gene Symbol: AKIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206646.1 | NP_001193575.1 | 5 | 521 | ||
NM_001206647.1 | NP_001193576.1 | 5 | 521 | |||
NM_001206648.1 | NP_001193577.1 | 4 | 440 | |||
NM_020642.3 | NP_065693.2 | 6 | 602 | |||
NR_045417.1 | 5 | 784 | ||||
NR_045418.1 | 4 | 703 | ||||
XM_017018011.1 | XP_016873500.1 | 6 | 622 | |||
XM_017018012.1 | XP_016873501.1 | 4 | 2518 | |||
GenBank mRNA | AF493783.1 | - | 6 | 602 | ||
AF493784.1 | - | 5 | 521 | |||
AF493785.1 | - | 5 | 521 | |||
AF512007.1 | - | 5 | 526 | |||
AK075308.1 | - | 721 | ||||
AW340643.1 | - | 3 | 295 | |||
BC030996.2 | - | 5 | 532 | 4609949 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
