siRNA Details
Gene Symbol: ABHD17B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016014.3 | NP_057098.2 | 5 | 1515 | ||
XM_006717134.3 | XP_006717197.2 | 5 | 966 | |||
XM_017014787.1 | XP_016870276.1 | 5 | 1122 | |||
XM_017014790.1 | XP_016870279.1 | 4 | 1323 | |||
GenBank mRNA | AF151825.1 | - | 5 | 1199 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
