siRNA Details
Gene Symbol: OR10P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206899.1 | NP_996782.1 | 1 | 874 | ||
GenBank mRNA | BC136914.1 | - | 1 | 906 | 9020905 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
