siRNA Details
Gene Symbol: LOC101927789 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005274417.3 | XP_005274474.1 | 1, 2 | 935 | ||
XM_011545424.2 | XP_011543726.1 | 1, 2 | 935 | |||
XM_017018642.1 | XP_016874131.1 | 1, 2 | 935 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
