siRNA Details
Gene Symbol: NDST2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003635.3 | NP_003626.1 | 1 | 71 | ||
XM_005270255.3 | XP_005270312.1 | 1 | 71 | |||
XM_017016857.1 | XP_016872346.1 | 1 | 71 | |||
GenBank mRNA | AK303648.1 | - | 1 | 71 | ||
BC018681.1 | - | 1 | 27 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
