siRNA Details
Gene Symbol: EPS8L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017729.3 | NP_060199.3 | 15 | 1887 | ||
NM_133180.2 | NP_573441.2 | 20 | 2270 | |||
XM_005259020.1 | XP_005259077.1 | 19 | 2366 | |||
XM_011527050.1 | XP_011525352.1 | 14 | 1983 | |||
GenBank mRNA | AF282167.1 | - | 10 | 1514 | ||
AF370395.1 | - | 20 | 2222 | |||
AK000265.1 | - | 15 | 1887 | |||
AK057052.1 | - | 10 | 1663 | |||
AK075098.1 | - | 20 | 2221 | |||
AK296732.1 | - | 18 | 2120 | |||
AK301615.1 | - | 9 | 1326 | |||
AY074928.1 | - | 19 | 2223 | |||
BC004907.2 | - | 12 | 1828 | 3529476 | ||
BC015763.1 | - | 15 | 1856 | 4871012 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
