siRNA Details
Gene Symbol: EPS8L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017729.3 | NP_060199.3 | 6, 7 | 791 | ||
NM_133180.2 | NP_573441.2 | 11, 12 | 1174 | |||
XM_005259020.1 | XP_005259077.1 | 11, 12 | 1174 | |||
XM_011527050.1 | XP_011525352.1 | 6, 7 | 791 | |||
XM_011527051.2 | XP_011525353.1 | 11, 12 | 1174 | |||
XM_011527052.2 | XP_011525354.1 | 11, 12 | 1174 | |||
GenBank mRNA | AF282167.1 | - | 2, 3 | 323 | ||
AF370395.1 | - | 11, 12 | 1126 | |||
AK000265.1 | - | 6, 7 | 791 | |||
AK057052.1 | - | 2, 3 | 471 | |||
AK075098.1 | - | 11, 12 | 1125 | |||
AK296732.1 | - | 9, 10 | 1024 | |||
AK309305.1 | - | 11, 12 | 1174 | |||
AK309908.1 | - | 9, 10 | 1100 | |||
AY074928.1 | - | 10, 11 | 1127 | |||
BC004907.2 | - | 8, 9 | 1194 | 3529476 | ||
BC015763.1 | - | 6, 7 | 760 | 4871012 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
