siRNA Details
Gene Symbol: WNT5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030775.2 | NP_110402.2 | 5 | 2174 | ||
NM_032642.2 | NP_116031.1 | 5 | 2212 | |||
GenBank mRNA | BC001749.1 | - | 5 | 2213 | 3528819 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
