siRNA Details
Gene Symbol: LMNA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282625.1 | NP_001269554.1 | 4 | 1001 | ||
NM_001282626.1 | NP_001269555.1 | 1 | 577 | |||
NM_005572.3 | NP_005563.1 | 1 | 577 | |||
NM_170707.3 | NP_733821.1 | 1 | 577 | |||
NM_170708.3 | NP_733822.1 | 1 | 577 | |||
GenBank mRNA | AK026584.1 | - | 1 | 533 | ||
AK056143.1 | - | 4 | 969 | |||
AK057997.1 | - | 1 | 426 | |||
AK122732.1 | - | 2 | 463 | |||
AK130179.1 | - | 1 | 484 | |||
AY357727.1 | - | 1 | 525 | |||
AY528714.1 | - | 1 | 446 | |||
AY847595.1 | - | 1 | 328 | |||
AY847596.1 | - | 1 | 328 | |||
AY847597.1 | - | 1 | 328 | |||
BC000511.2 | - | 1 | 508 | 2822703 | ||
BC003162.1 | - | 1 | 493 | 2958052 | ||
BC014507.1 | - | 1 | 525 | 4863480 | ||
BC033088.1 | - | 1 | 521 | 3623265 | ||
BG822820.1 | - | 2 | 602 | |||
M13451.1 | - | 1 | 462 | |||
X03444.1 | - | 1 | 538 | |||
X03445.1 | - | 1 | 538 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
