siRNA Details
Gene Symbol: LMNA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257374.2 | NP_001244303.1 | 4 | 435 | ||
NM_001282624.1 | NP_001269553.1 | 5 | 615 | |||
NM_001282625.1 | NP_001269554.1 | 7 | 1357 | |||
NM_001282626.1 | NP_001269555.1 | 4 | 933 | |||
NM_005572.3 | NP_005563.1 | 4 | 933 | |||
NM_170707.3 | NP_733821.1 | 4 | 933 | |||
NM_170708.3 | NP_733822.1 | 4 | 933 | |||
XM_011509533.1 | XP_011507835.1 | 4 | 430 | |||
XM_011509534.1 | XP_011507836.1 | 3 | 328 | |||
GenBank mRNA | AK026584.1 | - | 4 | 889 | ||
AK056191.1 | - | 5 | 508 | |||
AK057997.1 | - | 4 | 782 | |||
AK097801.1 | - | 5 | 543 | |||
AK098128.1 | - | 1 | 29 | |||
AK122732.1 | - | 5 | 819 | |||
AK130179.1 | - | 4 | 840 | |||
AK295390.1 | - | 4 | 382 | |||
AY357727.1 | - | 4 | 881 | |||
AY528714.1 | - | 4 | 802 | |||
AY847595.1 | - | 4 | 684 | |||
AY847596.1 | - | 4 | 684 | |||
AY847597.1 | - | 4 | 684 | |||
BC000511.2 | - | 4 | 864 | 2822703 | ||
BC003162.1 | - | 4 | 849 | 2958052 | ||
BC014507.1 | - | 4 | 881 | 4863480 | ||
BC033088.1 | - | 4 | 877 | 3623265 | ||
DC400775.1 | - | 4 | 392 | |||
HY027676.1 | - | 4 | 435 | |||
M13451.1 | - | 4 | 818 | |||
M13452.1 | - | 3 | 237 | |||
X03444.1 | - | 4 | 894 | |||
X03445.1 | - | 4 | 894 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
