siRNA Details
Gene Symbol: ZWINT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005413.1 | NP_001005413.1 | 4 | 360 | ||
NM_007057.3 | NP_008988.2 | 4 | 360 | |||
NM_032997.2 | NP_127490.1 | 4 | 360 | |||
XM_017015605.1 | XP_016871094.1 | 4 | 389 | |||
GenBank mRNA | AF067656.1 | - | 4 | 347 | ||
AK093808.1 | - | 2 | 661 | |||
AK225509.1 | - | 4 | 346 | |||
AK293416.1 | - | 4 | 349 | |||
AK298148.1 | - | 4 | 356 | |||
BC020979.1 | - | 4 | 347 | 3848873 | ||
BC110399.1 | - | 4 | 359 | 5554405 | ||
BG615304.1 | - | 4 | 360 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
