siRNA Details
Gene Symbol: ZWINT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005413.1 | NP_001005413.1 | 7, 8 | 695 | ||
NM_007057.3 | NP_008988.2 | 7, 8 | 836 | |||
NM_032997.2 | NP_127490.1 | 7, 8 | 836 | |||
XM_017015605.1 | XP_016871094.1 | 7, 8 | 828 | |||
GenBank mRNA | AF067656.1 | - | 7, 8 | 823 | ||
AK093808.1 | - | 5, 6 | 1137 | |||
AK225509.1 | - | 7, 8 | 822 | |||
AK293416.1 | - | 7, 8 | 825 | |||
AK298148.1 | - | 7, 8 | 691 | |||
BC000411.1 | - | 4, 5 | 448 | |||
BC020979.1 | - | 7, 8 | 823 | 3848873 | ||
BC110399.1 | - | 7, 8 | 835 | 5554405 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
